missing translation for 'onlineSavingsMsg'
Learn More

4,5-Dimethoxy-2-nitrobenzoic acid, 98%

Product Code. 11358205
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11358205 5 g 5g
11368205 25 g 25g
11378205 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11358205 Supplier Thermo Scientific Alfa Aesar Supplier No. L08510.06
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 4998-07-6 | C9H8NO6 | 226.17 g/mol

4,5-Dimethoxy-2-nitrobenzoic acid was used in the synthesis of 4,5-dimethoxy-2-nitrobenzamide and 6,7-dimethoxyquinazoline derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,5-Dimethoxy-2-nitrobenzoic acid was used in the synthesis of 4,5-dimethoxy-2-nitrobenzamide and 6,7-dimethoxyquinazoline derivatives.

Solubility
Insoluble in water. Soluble in DMSO and methanol. Solubility in hot 1mol/L NaOH (almost transparency).

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4998-07-6
MDL Number MFCD00014697

Specifications

Melting Point 192°C to 195°C
Molecular Formula C9H8NO6
Quantity 5 g
Beilstein 2123903
Synonym 4,5-dimethoxy-2-nitrobenzoic acid, 6-nitroveratric acid, 3,4-dimethoxy-6-nitrobenzoic acid, benzoic acid, 4,5-dimethoxy-2-nitro, zlchem 681, acmc-2097gn, ksc496i5r, 4,5-dimethoxy-2-nitrobenzoicacid, 2-nitro-4,5-dimethoxybenzoic acid, 4,5-dimethoxy-2-nitro-benzoic acid
Solubility Information Insoluble in water. Soluble in DMSO and methanol. Solubility in hot 1mol/L NaOH (almost transparency).
InChI Key WWCMFGBGMJAJRX-UHFFFAOYSA-M
SMILES COC1=C(C=C(C(=C1)C(=O)O)[N+](=O)[O-])OC
Molecular Weight (g/mol) 226.17
PubChem CID 78690
Formula Weight 227.17
Percent Purity 98%
Chemical Name or Material 4,5-Dimethoxy-2-nitrobenzoic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.