missing translation for 'onlineSavingsMsg'
Learn More

4,5-Dimethoxy-2-nitrobenzoic acid, 99%, Thermo Scientific™

Product Code. 15392022 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
Unit Size:
100g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15392022 100 g 100g
15372022 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15392022 Supplier Thermo Scientific Acros Supplier No. 334691000
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 4998-07-6
Molecular Formula C9H9NO6
Molecular Weight (g/mol) 227.17
InChI Key WWCMFGBGMJAJRX-UHFFFAOYSA-M
Synonym 4,5-dimethoxy-2-nitrobenzoic acid, 6-nitroveratric acid, 3,4-dimethoxy-6-nitrobenzoic acid, benzoic acid, 4,5-dimethoxy-2-nitro, zlchem 681, acmc-2097gn, ksc496i5r, 4,5-dimethoxy-2-nitrobenzoicacid, 2-nitro-4,5-dimethoxybenzoic acid, 4,5-dimethoxy-2-nitro-benzoic acid
PubChem CID 78690
SMILES COC1=C(C=C(C(=C1)C(=O)O)[N+](=O)[O-])OC

Specifications

Melting Point 194.0°C to 195.0°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Quantity 100 g
Formula Weight 227.17
Percent Purity 99%
Chemical Name or Material 4,5-Dimethoxy-2-nitrobenzoic acid
compliance-icons
Product Identifier
  • 4,5-Dimethoxy-2-nitrobenzoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.