missing translation for 'onlineSavingsMsg'
Learn More

4,5-Dimethoxy-2-nitrobenzoic acid, 99%, Thermo Scientific™

Product Code. 15382022 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15382022 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15382022 Supplier Thermo Scientific Acros Supplier No. 334690250
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 4998-07-6
CAS Min % 97.5
CAS Max % 100.0
Melting Point 194°C to 195°C
Color Yellow
Infrared Spectrum Authentic
Packaging Glass Bottle
Molecular Formula C9H9NO6
MDL Number MFCD00014697
Quantity 25 g
Synonym 4,5-dimethoxy-2-nitrobenzoic acid, 6-nitroveratric acid, 3,4-dimethoxy-6-nitrobenzoic acid, benzoic acid, 4,5-dimethoxy-2-nitro, zlchem 681, acmc-2097gn, ksc496i5r, 4,5-dimethoxy-2-nitrobenzoicacid, 2-nitro-4,5-dimethoxybenzoic acid, 4,5-dimethoxy-2-nitro-benzoic acid
InChI Key WWCMFGBGMJAJRX-UHFFFAOYSA-M
Molecular Weight (g/mol) 227.17
PubChem CID 78690
Formula Weight 227.17
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 4, 5-Dimethoxy-2-nitrobenzoic acid, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 4,5-Dimethoxy-2-nitrobenzoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.