missing translation for 'onlineSavingsMsg'
Learn More

4,5-Dimethoxy-2-nitrobenzyl alcohol, 98%

Product Code. 11330184
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11330184

Brand: Thermo Scientific Alfa Aesar L00719.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1016-58-6 | C9H11NO5 | 213.189 g/mol

4,5-Dimethoxy-2-nitrobenzyl alcohol, is used as Reagent for the protection of alcohols, etc., as ethers which can be cleaved by photolysis involving intramolecular rearrangement to derivatives of o-nitrosobenzaldehyde, which are readily cleaved by hydrolysis. It is also used as photochemical protecting groups in organic synthesis. It is also used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,5-Dimethoxy-2-nitrobenzyl alcohol, is used as Reagent for the protection of alcohols, etc., as ethers which can be cleaved by photolysis involving intramolecular rearrangement to derivatives of o-nitrosobenzaldehyde, which are readily cleaved by hydrolysis. It is also used as photochemical protecting groups in organic synthesis. It is also used as a pharmaceutical intermediate.

Solubility
Soluble in water.

Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1016-58-6
Molecular Formula C9H11NO5
Molecular Weight (g/mol) 213.189
MDL Number MFCD00014701
InChI Key WBSCOJBVYHQOFB-UHFFFAOYSA-N
Synonym 4,5-dimethoxy-2-nitrobenzyl alcohol, 6-nitroveratryl alcohol, 4,5-dimethoxy-2-nitrophenyl methanol, benzenemethanol, 4,5-dimethoxy-2-nitro, 2-nitro-4,5-dimethoxybenzyl alcohol, 3,4-dimethoxy-6-nitrobenzyl alcohol, 4,5-dimethoxy-2-nitrophenyl methan-1-ol, 6-nitroveratrylalcohol, pubchem8715, 4,5-dimethoxy 2-nitrobenzyl alcohol
PubChem CID 66097
IUPAC Name (4,5-dimethoxy-2-nitrophenyl)methanol
SMILES COC1=C(C=C(C(=C1)CO)[N+](=O)[O-])OC

Specifications

Melting Point 144°C to 148°C
Quantity 5 g
Beilstein 1880093
Sensitivity Light sensitive
Solubility Information Soluble in water.
Formula Weight 213.19
Percent Purity 98%
Chemical Name or Material 4,5-Dimethoxy-2-nitrobenzyl alcohol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.