missing translation for 'onlineSavingsMsg'
Learn More

4,5-Dimethoxy-2-nitrobenzyl alcohol, 98%

Product Code. 11340184
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11340184 25 g 25g
11330184 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11340184 Supplier Thermo Scientific Alfa Aesar Supplier No. L00719.14
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1016-58-6 | C9H11NO5 | 213.189 g/mol

4,5-Dimethoxy-2-nitrobenzyl alcohol, is used as Reagent for the protection of alcohols, etc., as ethers which can be cleaved by photolysis involving intramolecular rearrangement to derivatives of o-nitrosobenzaldehyde, which are readily cleaved by hydrolysis. It is also used as photochemical protecting groups in organic synthesis. It is also used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,5-Dimethoxy-2-nitrobenzyl alcohol, is used as Reagent for the protection of alcohols, etc., as ethers which can be cleaved by photolysis involving intramolecular rearrangement to derivatives of o-nitrosobenzaldehyde, which are readily cleaved by hydrolysis. It is also used as photochemical protecting groups in organic synthesis. It is also used as a pharmaceutical intermediate.

Solubility
Soluble in water.

Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1016-58-6
Molecular Formula C9H11NO5
Molecular Weight (g/mol) 213.189
MDL Number MFCD00014701
InChI Key WBSCOJBVYHQOFB-UHFFFAOYSA-N
Synonym 4,5-dimethoxy-2-nitrobenzyl alcohol, 6-nitroveratryl alcohol, 4,5-dimethoxy-2-nitrophenyl methanol, benzenemethanol, 4,5-dimethoxy-2-nitro, 2-nitro-4,5-dimethoxybenzyl alcohol, 3,4-dimethoxy-6-nitrobenzyl alcohol, 4,5-dimethoxy-2-nitrophenyl methan-1-ol, 6-nitroveratrylalcohol, pubchem8715, 4,5-dimethoxy 2-nitrobenzyl alcohol
PubChem CID 66097
IUPAC Name (4,5-dimethoxy-2-nitrophenyl)methanol
SMILES COC1=C(C=C(C(=C1)CO)[N+](=O)[O-])OC

Specifications

Melting Point 144°C to 148°C
Quantity 25 g
Beilstein 1880093
Sensitivity Light sensitive
Solubility Information Soluble in water.
Formula Weight 213.19
Percent Purity 98%
Chemical Name or Material 4,5-Dimethoxy-2-nitrobenzyl alcohol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.