missing translation for 'onlineSavingsMsg'
Learn More

4,5-Dimethoxy-2-nitrophenylacetic acid, 97%

Product Code. 11453128
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11453128 1 g 1g
11463128 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11453128 Supplier Thermo Scientific Alfa Aesar Supplier No. A18884.03
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 73357-18-3 | C10H11NO6 | 241.199 g/mol

It is employed as a intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a intermediate for pharmaceutical.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 73357-18-3
Molecular Formula C10H11NO6
Molecular Weight (g/mol) 241.199
MDL Number MFCD00118466
InChI Key WJPMJFUEMVXUCV-UHFFFAOYSA-N
Synonym 2-4,5-dimethoxy-2-nitrophenyl acetic acid, 4,5-dimethoxy-2-nitrophenyl acetic acid, 4,5-dimethoxy-2-nitrophenylacetic acid, 4,5-dimethoxy-2-nitro-phenyl-acetic acid, 4,5-dimethoxy-2-nitro-phenylacetic acid, benzeneacetic acid, 4,5-dimethoxy-2-nitro, maybridge1_002500, acmc-1bm2u, cbmicro_008990, cambridge id 5234450
PubChem CID 270941
IUPAC Name 2-(4,5-dimethoxy-2-nitrophenyl)acetic acid
SMILES COC1=C(C=C(C(=C1)CC(=O)O)[N+](=O)[O-])OC

Specifications

Melting Point 206°C to 208°C
Quantity 1 g
Formula Weight 241.2
Percent Purity 97%
Chemical Name or Material 4,5-Dimethoxy-2-nitrophenylacetic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.