missing translation for 'onlineSavingsMsg'
Learn More

4,5-Dimethoxy-2-nitrophenylacetonitrile, 98%

Product Code. 15455507
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15455507 250 mg 250mg
15445507 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15455507 Supplier Thermo Scientific Alfa Aesar Supplier No. H32747.MD
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 17354-04-0 | C10H10N2O4 | 222.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17354-04-0
Molecular Formula C10H10N2O4
Molecular Weight (g/mol) 222.20
MDL Number MFCD00033967
InChI Key PKDJSFDIQCAPSY-UHFFFAOYSA-N
Synonym 2-4,5-dimethoxy-2-nitrophenyl acetonitrile, 4,5-dimethoxy-2-nitrophenyl acetonitrile, 4,5-dimethoxy-2-nitrophenylacetonitrile, benzeneacetonitrile,4,5-dimethoxy-2-nitro, benzeneacetonitrile, 4,5-dimethoxy-2-nitro, acmc-1bvbg, 2-nitro-4,5-dimethoxybenzyl cyanide, 2-4,5-dimethoxy-2-nitrophenyl ethanenitrile, 4,5-dimethoxy-2-nitro-phenyl-acetonitrile
PubChem CID 222992
IUPAC Name 2-(4,5-dimethoxy-2-nitrophenyl)acetonitrile
SMILES COC1=CC(CC#N)=C(C=C1OC)[N+]([O-])=O

Specifications

Melting Point 110°C to 112°C
Quantity 250 mg
UN Number UN3439
Formula Weight 222.2
Percent Purity 98%
Chemical Name or Material 4,5-Dimethoxy-2-nitrophenylacetonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.