missing translation for 'onlineSavingsMsg'
Learn More

4-[(6-Methylpyrazin-2-yl)oxy]aniline, 97%, Thermo Scientific™

Product Code. 10143624
Change view
Click to view available options
Amber Glass Bottle; 250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10143624 Amber Glass Bottle; 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10143624 Supplier Thermo Scientific Maybridge Supplier No. CC62714CB
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
This item is not returnable. View return policy

Specifications

CAS 915707-63-0
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C11H11N3O
MDL Number MFCD09702409
Quantity 250 mg
Synonym 4-6-methylpyrazin-2-yl oxy aniline, 4-6-methylpyrazin-2-yloxy aniline, 2-4-aminophenoxy-6-methylpyrazine, 4-6-methylpyrazin-2-yloxy phenylamine, benzenamine,4-6-methyl-2-pyrazinyl oxy
InChI Key OZPUGVATAOJVLW-UHFFFAOYSA-N
SMILES CC1=CN=CC(=N1)OC2=CC=C(C=C2)N
IUPAC Name 4-(6-methylpyrazin-2-yl)oxyaniline
Molecular Weight (g/mol) 201.229
PubChem CID 24229694
Formula Weight 201.23
Percent Purity 97%
Chemical Name or Material 4-[(6-Methylpyrazin-2-yl)oxy]aniline
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.