missing translation for 'onlineSavingsMsg'
Learn More

4',7-Dihydroxyisoflavone, 97%

Product Code. 11439927
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11439927 1 g 1g
11449927 5 g 5g
11459927 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11439927 Supplier Thermo Scientific Alfa Aesar Supplier No. B22877.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 486-66-8 | C15H10O4 | 254.241 g/mol

4',7-Dihydroxyisoflavone is a phytoestrogen that is suggested to play a role in preventing hormone-induced cancers. It protects against oxidative damage in liver cells induced by 7,12-dimethylbenz[a]anthracene (DMBA). Catalase and superoxide dismutase activity, down-regulated by DMBA, was restored by daidzein.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4′,7-Dihydroxyisoflavone is a phytoestrogen that is suggested to play a role in preventing hormone-induced cancers. It protects against oxidative damage in liver cells induced by 7,12-dimethylbenz[a]anthracene (DMBA). Catalase and superoxide dismutase activity, down-regulated by DMBA, was restored by daidzein.

Solubility
Insoluble in water. Soluble in DMSO (25 mg/ml), ethanol (2 mg/ml), DMF (10 mg/ml), and 1:10 DMSO:PBS (pH 7.2) (0.15 mg/ml).

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 486-66-8
Molecular Formula C15H10O4
Molecular Weight (g/mol) 254.241
MDL Number MFCD00016954
InChI Key ZQSIJRDFPHDXIC-UHFFFAOYSA-N
Synonym daidzein, 4',7-dihydroxyisoflavone, daidzeol, 7,4'-dihydroxyisoflavone, 7-hydroxy-3-4-hydroxyphenyl-4h-chromen-4-one, diadzein, 7-hydroxy-3-4-hydroxyphenyl-4h-1-benzopyran-4-one, 4h-1-benzopyran-4-one, 7-hydroxy-3-4-hydroxyphenyl, 7-hydroxy-3-4-hydroxyphenyl chromen-4-one, 7-hydroxy-3-4-hydroxyphenyl-4-benzopyrone
PubChem CID 5281708
ChEBI CHEBI:28197
IUPAC Name 7-hydroxy-3-(4-hydroxyphenyl)chromen-4-one
SMILES C1=CC(=CC=C1C2=COC3=C(C2=O)C=CC(=C3)O)O

Specifications

Melting Point 328°C to 332°C (decomposition)
Quantity 1 g
Beilstein 231523
Merck Index 14,2801
Solubility Information Insoluble in water. Soluble in DMSO (25mg/ml),ethanol (2mg/ml),DMF (10mg/ml),and 1:10 DMSO:PBS (pH 7.2) (0.15mg/ml).
Formula Weight 254.24
Percent Purity 97%
Chemical Name or Material 4',7-Dihydroxyisoflavone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.