missing translation for 'onlineSavingsMsg'
Learn More

4-Acetamidobenzaldehyde, 98%

Product Code. 11454377
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11454377 5 g 5g
11464377 25 g 25g
11474377 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11454377 Supplier Thermo Scientific Alfa Aesar Supplier No. B20430.06
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 122-85-0 | C9H9NO2 | 163.176 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122-85-0
Molecular Formula C9H9NO2
Molecular Weight (g/mol) 163.176
MDL Number MFCD00003380
InChI Key SKLUWKYNZNXSLX-UHFFFAOYSA-N
Synonym 4-acetamidobenzaldehyde, n-4-formylphenyl acetamide, 4-acetylaminobenzaldehyde, p-formylacetanilide, acetamide, n-4-formylphenyl, micotiazone, 4-formylacetanilide, 4'-formylacetanilide, p-acetamidobenzaldehyde, p-acetaminobenzaldehyde
PubChem CID 73942
IUPAC Name N-(4-formylphenyl)acetamide
SMILES CC(=O)NC1=CC=C(C=C1)C=O

Specifications

Melting Point 154°C to 157°C
Quantity 5 g
Sensitivity Air Sensitive
Formula Weight 163.18
Percent Purity 98%
Chemical Name or Material 4-Acetamidobenzaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.