missing translation for 'onlineSavingsMsg'
Learn More

4-Acetamidophenylboronic acid, 97+%

Product Code. 10782993 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10782993 5 g Glass bottle 5g
10532242 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10782993 Supplier Thermo Scientific Acros Supplier No. 367610050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 101251-09-6 | C8H10BNO3 | 178.98 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 101251-09-6
Molecular Formula C8H10BNO3
Molecular Weight (g/mol) 178.98
MDL Number MFCD02179451
InChI Key VYEWTHXZHHATTA-UHFFFAOYSA-N
Synonym 4-acetylaminophenylboronic acid, 4-acetamidophenyl boronic acid, p-acetamidophenylboronic acid, 4-acetamidobenzeneboronic acid, 4-acetamidophenyl-boronic acid, 4-acetaminophenyl boronic acid, 4-acetylaminophenyl boronic acid, 4-acetylamino phenylboronic acid, boronic acid, 4-acetylamino phenyl
PubChem CID 2734657
IUPAC Name (4-acetamidophenyl)boronic acid
SMILES CC(=O)NC1=CC=C(C=C1)B(O)O

Specifications

CAS Min % 97.0
CAS Max % 100.0
Melting Point 215.0°C to 220.0°C
Color Gray-Tan
Infrared Spectrum Authentic
Assay Percent Range 97% min. (GC)
Packaging Glass bottle
Quantity 5 g
Solubility Information Solubility in water: 85g/L (70°C). Other solubilities: slightly soluble in ethylacetate,soluble in meoh,etoh,dmso
Formula Weight 178.98
Percent Purity 97+%
Physical Form Powder
Chemical Name or Material 4-Acetamidophenylboronic acid
Show More Show Less
compliance-icons
Product Identifier
  • 4-Acetamidophenylboronic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.