missing translation for 'onlineSavingsMsg'
Learn More

4-Acetamidothiophenol, 95%, Thermo Scientific™

Product Code. 10152890 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10152890 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10152890 Supplier Thermo Scientific Acros Supplier No. 102340050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1126-81-4
Molecular Formula C8H9NOS
Molecular Weight (g/mol) 167.23
MDL Number MFCD00004848
InChI Key AYEQJLOHMLYKAV-UHFFFAOYSA-N
Synonym 4-acetamidothiophenol, 4'-mercaptoacetanilide, 4-acetamidobenzenethiol, n-4-sulfanylphenyl acetamide, 4-acetamido thiophenol, n-4-mercaptophenyl acetamide, acetanilide, 4'-mercapto, 4-acetaminothiophenol, 4-mercaptoacetanilide, 4-acetylaminothiophenol
PubChem CID 70777
IUPAC Name N-(4-sulfanylphenyl)acetamide
SMILES CC(=O)NC1=CC=C(S)C=C1

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 149°C to 154°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Linear Formula CH3CONHC6H4SH
Quantity 5 g
Beilstein 13, 541
Formula Weight 167.23
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 4-Acetamidothiophenol, 95%
Mehr anzeigen Weniger anzeigen
compliance-icons
Produktidentifikator
  • 4-Acetamidothiophenol
Signalwort
  • Warning
Gefahrenkategorie
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Gefahrenhinweis
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Sicherheitshinweis
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.