missing translation for 'onlineSavingsMsg'
Learn More

4-Acetophenetidide, 97%

Product Code. p-48703 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Unit Size:
100g
2.5kg
500g
This item is not returnable. View return policy

Product Code. 10062830

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 62-44-2 | C10H13NO2 | 179.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62-44-2
Molecular Formula C10H13NO2
Molecular Weight (g/mol) 179.22
MDL Number MFCD00009094
InChI Key CPJSUEIXXCENMM-UHFFFAOYSA-N
Synonym phenacetin, acetophenetidin, n-4-ethoxyphenyl acetamide, acetphenetidin, acetophenetidine, acetophenetin, phenacetine, p-acetophenetidide, contradouleur, achrocidin
PubChem CID 4754
ChEBI CHEBI:8050
IUPAC Name N-(4-ethoxyphenyl)acetamide
SMILES CCOC1=CC=C(C=C1)NC(=O)C

Specifications

Melting Point 133.0°C to 138.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Plastic bottle
Linear Formula CH3CONHC6H4OC2H5
Quantity 100 g
Beilstein 13, 461
Merck Index 15, 7319
Solubility Information Solubility in water: 1g/1310mL cold water. Other solubilities: 1g/82mL boiling water, 1g/15mL cold alcohol, 1g/2.8mL boiling alcohol, 1g/14mL chloroform, 1g/90mL ether - soluble in glycerol
Formula Weight 179.22
Percent Purity 97%
Physical Form Fine Crystalline Powder
Chemical Name or Material 4-Acetophenetidide, 97%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • p-Acetophenetidide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 1A
Hazard Statement
  • H350-May cause cancer.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.