missing translation for 'onlineSavingsMsg'
Learn More

4-Acetoxy-3-ethoxybenzaldehyde, 99%, Thermo Scientific™

Product Code. 11355559
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11355559 250 mg 250mg
11365559 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11355559 Supplier Thermo Scientific Alfa Aesar Supplier No. H50488.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
4-Acetoxy-3-ethoxybenzaldehyde is used in organic synthesis. It is used as catalytic agent; petrochemical additive.

Solubility
Soluble in water (986.4 mg / L) at 25°C.

Notes
Air Sensitive. Store in a cool, dry, well-ventilated area away from incompatible substances such as oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 72207-94-4
Molecular Formula C11H12O4
Molecular Weight (g/mol) 208.213
MDL Number MFCD01666413
InChI Key XRZFVPCFHPIMAD-UHFFFAOYSA-N
Synonym 4-acetoxy-3-ethoxybenzaldehyde, ethyl vanillin acetate, benzaldehyde, 4-acetyloxy-3-ethoxy, 4-formyl-2-ethoxyphenyl ethanoate, 2-ethoxy-4-formylphenyl acetate, 2-ethoxy-4-formyl-phenyl acetate, ethyl vanilline acetate, 3-08-00-02030 beilstein handbook reference, 4-acetyloxy-3-ethoxybenzaldehyde
PubChem CID 155708
IUPAC Name (2-ethoxy-4-formylphenyl) acetate
SMILES CCOC1=C(C=CC(=C1)C=O)OC(=O)C

Specifications

Quantity 250 mg
Sensitivity Air Sensitive
Solubility Information Soluble in water (986.4 mg / L) at 25°C.
Formula Weight 208.22
Percent Purity 99%
Chemical Name or Material 4-Acetoxy-3-ethoxybenzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.