missing translation for 'onlineSavingsMsg'
Learn More

4-Acetoxycinnamic acid, predominantly trans, 98+%

Product Code. 11429853
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11429853 100 g 100g
11409853 5 g 5g
11419853 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11429853 Supplier Thermo Scientific Alfa Aesar Supplier No. A12979.22
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 15486-19-8 | C11H10O4 | 206.197 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15486-19-8
Molecular Formula C11H10O4
Molecular Weight (g/mol) 206.197
MDL Number MFCD00016847
InChI Key BYHBHNKBISXCEP-QPJJXVBHSA-N
Synonym 4-acetoxycinnamic acid, p-acetoxycinnamic acid, 3-4-acetoxyphenyl acrylic acid, 2-propenoic acid, 3-4-acetyloxy phenyl, trans-4-acetoxycinnamic acid, 3-4-acetyloxy phenyl prop-2-enoic acid, 4-acetoxycinnamic acid, predominantly trans, acetylated coumaric acid, 2e-3-4-acetyloxy phenyl prop-2-enoic acid, 3-4-acetoxyphenyl-2-propenoic acid
PubChem CID 5373941
ChEBI CHEBI:86580
IUPAC Name (E)-3-(4-acetyloxyphenyl)prop-2-enoic acid
SMILES CC(=O)OC1=CC=C(C=C1)C=CC(=O)O

Specifications

Melting Point 205°C to 208°C
Odor Odorless
Quantity 100 g
Beilstein 2645987
Formula Weight 206.2
Percent Purity ≥98%
Chemical Name or Material 4-Acetoxycinnamic acid, predominantly trans

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.