missing translation for 'onlineSavingsMsg'
Learn More

4-Allylanisole, 98%

Product Code. 10765931 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10765931 25 g Glass bottle 25g
10570692 5 g Glass bottle 5g
10030570 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10765931 Supplier Thermo Scientific Acros Supplier No. 102880250
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 140-67-0 | C10H12O | 148.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 140-67-0
Molecular Formula C10H12O
Molecular Weight (g/mol) 148.2
MDL Number MFCD00008653
InChI Key ZFMSMUAANRJZFM-UHFFFAOYSA-N
Synonym estragole, 4-allylanisole, p-allylanisole, tarragon, methyl chavicol, 1-allyl-4-methoxybenzene, estragol, esdragol, esdragole, esdragon
PubChem CID 8815
ChEBI CHEBI:4867
IUPAC Name 1-methoxy-4-prop-2-enylbenzene
SMILES COC1=CC=C(C=C1)CC=C

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless
Density 0.9600g/mL
Boiling Point 215°C to 216°C
Flash Point 81°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula H2C=CHCH2C6H4OCH3
Refractive Index 1.5205 to 1.5225
Quantity 25 g
Beilstein 06, 571
Merck Index 15, 3760
Specific Gravity 0.96
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol and chloroform
Formula Weight 148.2
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 4-Allylanisole, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Allylanisole
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.