missing translation for 'onlineSavingsMsg'
Learn More

4-Allylanisole, 98%

Product Code. 11470917
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11470917 100 g 100g
11460917 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11470917 Supplier Thermo Scientific Alfa Aesar Supplier No. B23311.22
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 140-67-0 | C10H12O | 148.205 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 140-67-0
Molecular Formula C10H12O
Molecular Weight (g/mol) 148.205
MDL Number MFCD00008653
InChI Key ZFMSMUAANRJZFM-UHFFFAOYSA-N
Synonym estragole, 4-allylanisole, p-allylanisole, tarragon, methyl chavicol, 1-allyl-4-methoxybenzene, estragol, esdragol, esdragole, esdragon
PubChem CID 8815
ChEBI CHEBI:4867
IUPAC Name 1-methoxy-4-prop-2-enylbenzene
SMILES COC1=CC=C(C=C1)CC=C

Specifications

Density 0.965
Boiling Point 215°C to 216°C
Flash Point 81°C (177°F)
Refractive Index 1.521
Quantity 100 g
Beilstein 1099454
Merck Index 14,3705
Formula Weight 148.21
Percent Purity 98%
Chemical Name or Material 4-Allylanisole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.