missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-2,3,5,6-tetrafluorobenzoic acid, 97%

Product Code. 11314576
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11314576 1 g 1g
11324576 5 g 5g
11334576 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11314576 Supplier Thermo Scientific Alfa Aesar Supplier No. L20128.03
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 944-43-4 | C7H3F4NO2 | 209.1 g/mol

It is used in chemical research and as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in chemical research and as pharmaceutical intermediate.

Solubility
Insoluble in water. Soluble in methanol (25 mg/ml).

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 944-43-4
Molecular Formula C7H3F4NO2
Molecular Weight (g/mol) 209.1
MDL Number MFCD00007647
InChI Key WTNSXWSOTDBWOR-UHFFFAOYSA-N
Synonym 2,3,5,6-tetrafluoro-4-aminobenzoic acid, acmc-209rrw, 4-amino-2,3,5,6-tetrafluorobenzoicacid, 2,3,5,6-tetrafluoro-4-amino-benzoic acid, 4-amino-2,3,5,6-tetrafluoro-benzoic acid, benzoic acid,4-amino-2,3,5,6-tetrafluoro, benzoic acid, 4-amino-2,3,5,6-tetrafluoro, 4-amino-2,3,5,6-tetrafluorobenzoic acid
PubChem CID 70345
IUPAC Name 4-amino-2,3,5,6-tetrafluorobenzoic acid
SMILES C1(=C(C(=C(C(=C1F)F)N)F)F)C(=O)O

Specifications

Melting Point 182°C to 184°C
Quantity 1 g
Beilstein 2121117
Solubility Information Insoluble in water. Soluble in methanol (25mg/ml).
Formula Weight 209.11
Percent Purity 98%
Chemical Name or Material 4-Amino-2,3,5,6-tetrafluorobenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.