missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-2-chlorobenzoic acid, 97%

Product Code. 10750901 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10750901 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10750901 Supplier Thermo Scientific Acros Supplier No. 203230050
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 2457-76-3 | C7H6ClNO2 | 171.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 2457-76-3
CAS Min % 96.0
CAS Max % 100.0
Melting Point 210°C to 215°C
Color Beige to Brown
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Molecular Formula C7H6ClNO2
Linear Formula H2NC6H3(Cl)CO2H
MDL Number MFCD00007772
Quantity 5 g
Beilstein 14, 438
Synonym 2-chloro-4-aminobenzoic acid, benzoic acid, 4-amino-2-chloro, o-chloro-p-aminobenzoic acid, 2-chloro-p-aminobenzoic acid, usaf nb-1, 4-amino-2-chloro-benzoic acid, 4-amino-2-chlorobenzoicacid, unii-y3s6924ia6, wln: zr cg dvq, pubchem3109
InChI Key MBDUKNCPOPMRJQ-UHFFFAOYSA-N
SMILES C1=CC(=C(C=C1N)Cl)C(=O)O
IUPAC Name 4-amino-2-chlorobenzoic acid
Molecular Weight (g/mol) 171.58
PubChem CID 17154
ChEBI CHEBI:59472
Formula Weight 171.58
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 4-Amino-2-chlorobenzoic acid, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Amino-2-chlorobenzoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.