missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-2-fluoro-5-methoxybenzeneboronic acid pinacol ester, 96%, Thermo Scientific Chemicals

Product Code. 15455366
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15455366 1 g 1g
15465366 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15455366 Supplier Thermo Scientific Alfa Aesar Supplier No. H59944.03
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket

missing translation for 'validMainframeMsg'

Denna artikel kan inte returneras. Se returpolicy

CAS: 1326283-60-6 | Molecular Formula: C13H19BFNO3 | Molecular Weight: 267.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 1326283-60-6
Molecular Formula C13H19BFNO3
Molecular Weight (g/mol) 267.11
MDL Number MFCD22419265
InChI Key JWACNMHKJHEWAQ-UHFFFAOYSA-N
Synonym 4-amino-2-fluoro-5-methoxybenzeneboronic acid pinacol ester, 5-fluoro-2-methoxy-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline
PubChem CID 67363295
IUPAC Name 5-fluoro-2-methoxy-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
SMILES COC1=C(N)C=C(F)C(=C1)B1OC(C)(C)C(C)(C)O1

Specifikationer

Quantity 1 g
Formula Weight 267.11
Percent Purity 96%
Chemical Name or Material 4-Amino-2-fluoro-5-methoxybenzeneboronic acid pinacol ester

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.