missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-2-methylbenzoic acid, 98%

Product Code. 15424267
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15424267 1 g 1g
15434267 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15424267 Supplier Thermo Scientific Alfa Aesar Supplier No. H31657.03
 Restricted Item
Estimated Shipment: 22-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 2486-75-1 | C8H9NO2 | 151.165 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2486-75-1
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.165
MDL Number MFCD06656133
InChI Key XRSQZFJLEPBPOZ-UHFFFAOYSA-N
Synonym 4-amino-2-methyl-benzoic acid, 2-methyl-4-aminobenzoic acid, 2-methyl-4-amino-benzoic acid, 4-amino-2-methyl benzoic acid, benzoic acid, 4-amino-2-methyl, pubchem10924, 4-amino-o-toluic acid, methyl-p-aminobenzoesaure, acmc-1cnb3, methyl-4-amino benzoic acid
PubChem CID 241632
IUPAC Name 4-amino-2-methylbenzoic acid
SMILES CC1=C(C=CC(=C1)N)C(=O)O

Specifications

Quantity 1 g
Formula Weight 151.16
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 4-Amino-2-methylbenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.