missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-2-(trifluoromethyl)benzonitrile, 97%

Product Code. 11945641 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11945641 1 g 1g
11955641 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11945641 Supplier Thermo Scientific Acros Supplier No. 442470010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 654-70-6 | C8H5F3N2 | 186.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 654-70-6
Molecular Formula C8H5F3N2
Molecular Weight (g/mol) 186.14
InChI Key PMDYLCUKSLBUHO-UHFFFAOYSA-N
Synonym 4-amino-2-trifluoromethyl benzonitrile, 5-amino-2-cyanobenzotrifluoride, 4-cyano-3-trifluoromethylaniline, 2-cyano-5-aminobenzotrifluoride, 4-cyano-3-trifluoromethyl aniline, benzonitrile, 4-amino-2-trifluoromethyl, unii-l47d9xhc08, 4-amino 2-trifluoromethyl benzonitrile, 2-trifluoromethyl-4-aminobenzonitrile, 4-amino-2-trifluoromethyl benzenecarbonitrile
PubChem CID 522170
IUPAC Name 4-amino-2-(trifluoromethyl)benzonitrile
SMILES C1=CC(=C(C=C1N)C(F)(F)F)C#N

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 144.0°C to 146.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 186.14
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 4-Amino-2-(trifluoromethyl)benzonitrile
Show More Show Less
compliance-icons
Product Identifier
  • 4-Amino-2-(trifluoromethyl)benzonitrile
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.