missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-3,5-dichlorobenzoic acid, 98%

Product Code. p-7022282
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11437197

Brand: Thermo Scientific Alfa Aesar A14982.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 56961-25-2 | C7H5Cl2NO2 | 206.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56961-25-2
Molecular Formula C7H5Cl2NO2
Molecular Weight (g/mol) 206.02
MDL Number MFCD00017091
InChI Key UHXYYTSWBYTDPD-UHFFFAOYSA-N
Synonym 3,5-dichloro-4-aminobenzoic acid, unii-bik7ns3b2f, benzoic acid, 4-amino-3,5-dichloro, bik7ns3b2f, 4-amino-3,5-dichlorobenzoicacid, pubchem14451, acmc-209xmx, 4-14-00-01279 beilstein handbook reference, ksc495s0n, 4-carboxy-2,6-dichloroaniline
PubChem CID 42061
IUPAC Name 4-amino-3,5-dichlorobenzoic acid
SMILES NC1=C(Cl)C=C(C=C1Cl)C(O)=O

Specifications

Melting Point ∼290°C (decomposition)
Quantity 5 g
Beilstein 2805751
Formula Weight 206.03
Percent Purity 98%
Chemical Name or Material 4-Amino-3,5-dichlorobenzoic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.