missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-3,5-diiodobenzoic acid, tech. 90%

Product Code. 11413324
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11413324 25 g 25g
11403324 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11413324 Supplier Thermo Scientific Alfa Aesar Supplier No. A10617.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 2122-61-4 | C7H5I2NO2 | 388.93 g/mol

4-Amino-3,5-diiodobenzoic acid is a very useful substituted benzene compound for use as a building block in synthesis. The carboxylic acid and amine functionalities can be used to form linkages through nucleophile-electrophile coupling and the iodides may be used as functional sites for cross-coupling transformations. 4-Amino-3,5-diiodobenzoic acid has been specifically used in the synthesis of an adduct to introduce into Tecoflex polymers to impart radiopaqueness to the material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Amino-3,5-diiodobenzoic acid is a very useful substituted benzene compound for use as a building block in synthesis. The carboxylic acid and amine functionalities can be used to form linkages through nucleophile-electrophile coupling and the iodides may be used as functional sites for cross-coupling transformations. 4-Amino-3,5-diiodobenzoic acid has been specifically used in the synthesis of an adduct to introduce into Tecoflex polymers to impart radiopaqueness to the material.

Solubility
Soluble in water (partly miscible).

Notes
Store at room temperature. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents. Stable under normal temperatures and pressures.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2122-61-4
Molecular Formula C7H5I2NO2
Molecular Weight (g/mol) 388.93
MDL Number MFCD00007684
InChI Key WXTVPMWCUMEVSZ-UHFFFAOYSA-N
Synonym benzoic acid, 4-amino-3,5-diiodo, 3,5-dijod-4-aminohippursaeure, 3,5-diiodo-4-aminobenzoic acid, 3,5-dijod-4-aminohippursaeure german, 4-amino-3,5-diiodo-benzoicacid, pubchem4681, acmc-1cq3z, wln: zr bi fi dvq, labotest-bb lt00080707, 4-amino-3,5-diiodo-benzoicaci
PubChem CID 16460
IUPAC Name 4-amino-3,5-diiodobenzoic acid
SMILES NC1=C(I)C=C(C=C1I)C(O)=O

Specifications

Melting Point >300°C
Quantity 25 g
Beilstein 2103037
Sensitivity Light sensitive
Solubility Information Soluble in water (partly miscible).
Formula Weight 388.93
Percent Purity 90%
Grade Technical
Chemical Name or Material 4-Amino-3,5-diiodobenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.