missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-3-bromobenzonitrile, 97%

Product Code. 15491748
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15491748 5 g 5g
15401758 250 mg 250mg
15481748 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15491748 Supplier Thermo Scientific Alfa Aesar Supplier No. H55483.06
 Restricted Item
Estimated Shipment: 21-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 50397-74-5 | C7H5BrN2 | 197.035 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50397-74-5
Molecular Formula C7H5BrN2
Molecular Weight (g/mol) 197.035
MDL Number MFCD01861386
InChI Key POESQIHWIIWNJL-UHFFFAOYSA-N
Synonym 3-bromo-4-aminobenzonitrile, 2-bromo-4-cyanoaniline, benzonitrile, 4-amino-3-bromo, 4-amino-3-bromo-benzonitrile, 4-amino-3-brombenzonitrile, buttpark 35\03-64, 4-amino-3-bromobenzenecarbonitrile, pubchem4609, acmc-209klz, 4-amino-3-bromobenzonitile
PubChem CID 1515279
IUPAC Name 4-amino-3-bromobenzonitrile
SMILES C1=CC(=C(C=C1C#N)Br)N

Specifications

Melting Point 106°C to 110°C
Quantity 5 g
UN Number UN3439
Formula Weight 197.03
Percent Purity 97%
Chemical Name or Material 4-Amino-3-bromobenzonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.