missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-3-fluorophenol, 96%

Product Code. p-7028680
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15423367

Brand: Thermo Scientific Alfa Aesar H26084.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 399-95-1 | C6H6FNO | 127.118 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 399-95-1
Molecular Formula C6H6FNO
Molecular Weight (g/mol) 127.118
MDL Number MFCD00077452
InChI Key MNPLTKHJEAFOCA-UHFFFAOYSA-N
Synonym 3-fluoro-4-aminophenol, 2-fluoro-4-hydroxyaniline, phenol, 4-amino-3-fluoro, 4-amino-3-fluoro-phenol, pubchem2832, 3-fluoro-4-amino-phenol, 4-amino-3-fluorophenol, acmc-209j9q, 4-amine-3-fluorophenol, buttpark 33\04-79
PubChem CID 2735919
IUPAC Name 4-amino-3-fluorophenol
SMILES C1=CC(=C(C=C1O)F)N

Specifications

Melting Point 137°C to 140°C
Quantity 250 mg
UN Number UN3077
Formula Weight 127.12
Percent Purity 96%
Chemical Name or Material 4-Amino-3-fluorophenol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.