missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-3-hydroxy-1-naphthalenesulfonic acid, 99%

Product Code. 11465767
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
Product Code. Quantity unitSize
11465767 25 g 25g
11475767 100 g 100g
2 options
This item is not returnable. View return policy

Product Code. 11465767

Brand: Thermo Scientific Alfa Aesar B24339.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 116-63-2 | C10H9NO4S | 239.245 g/mol

4-Amino-3-hydroxy-1-naphthalenesulfonic acid is used as mordant dyes and acid dyes intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Amino-3-hydroxy-1-naphthalenesulfonic acid is used as mordant dyes and acid dyes intermediate.

Solubility
Soluble in bases. Insoluble in water.

Notes
Store away from strong oxidizers, strong bases, acid anhydrides, acid chlorides.
TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 116-63-2
Molecular Formula C10H9NO4S
Molecular Weight (g/mol) 239.245
MDL Number MFCD00004019
InChI Key RXCMFQDTWCCLBL-UHFFFAOYSA-N
Synonym 1-amino-2-naphthol-4-sulfonic acid, 4-amino-3-hydroxy-1-naphthalenesulfonic acid, 1-amino-4-sulfo-2-naphthol, 1,2,4-acid, unii-w46sk3p33g, 1-naphthalenesulfonic acid, 4-amino-3-hydroxy, 4-amino-3-hydroxynaphthalene-1-sulphonic acid, 4-amino-3-hydroxynaphthalenesulfonic acid, 1,2,4-aminonaphtholsulfonic acid, 2-hydroxy-4-sulfo-1-naphthylamine
PubChem CID 8316
ChEBI CHEBI:19024
IUPAC Name 4-amino-3-hydroxynaphthalene-1-sulfonic acid
SMILES C1=CC=C2C(=C1)C(=CC(=C2N)O)S(=O)(=O)O

Specifica

Melting Point ∼295°C (decomposition)
Quantity 25 g
Beilstein 2697469
Merck Index 14,454
Solubility Information Soluble in bases. Insoluble in water.
Formula Weight 239.25
Percent Purity 99%
Chemical Name or Material 4-Amino-3-hydroxy-1-naphthalenesulfonic acid
compliance-icons
Identificativo prodotto
  • 4-Amino-3-hydroxy-1-naphthalenesulfonic acid
Avvertenza
  • Warning
Categoria di pericolo
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Indicazioni di pericolo
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Consigli di prudenza
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Informazioni supplementari
  • MIXTURE LIST-Contains : 4-Amino-3-hydroxy-1-naphthalenesulfonicacid

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.