missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-3-hydroxy-1-naphthalenesulfonic acid, 99%

Product Code. p-7027523
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11465767

Brand: Thermo Scientific Alfa Aesar B24339.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 116-63-2 | C10H9NO4S | 239.245 g/mol

4-Amino-3-hydroxy-1-naphthalenesulfonic acid is used as mordant dyes and acid dyes intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Amino-3-hydroxy-1-naphthalenesulfonic acid is used as mordant dyes and acid dyes intermediate.

Solubility
Soluble in bases. Insoluble in water.

Notes
Store away from strong oxidizers, strong bases, acid anhydrides, acid chlorides.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 116-63-2
Molecular Formula C10H9NO4S
Molecular Weight (g/mol) 239.245
MDL Number MFCD00004019
InChI Key RXCMFQDTWCCLBL-UHFFFAOYSA-N
Synonym 1-amino-2-naphthol-4-sulfonic acid, 4-amino-3-hydroxy-1-naphthalenesulfonic acid, 1-amino-4-sulfo-2-naphthol, 1,2,4-acid, unii-w46sk3p33g, 1-naphthalenesulfonic acid, 4-amino-3-hydroxy, 4-amino-3-hydroxynaphthalene-1-sulphonic acid, 4-amino-3-hydroxynaphthalenesulfonic acid, 1,2,4-aminonaphtholsulfonic acid, 2-hydroxy-4-sulfo-1-naphthylamine
PubChem CID 8316
ChEBI CHEBI:19024
IUPAC Name 4-amino-3-hydroxynaphthalene-1-sulfonic acid
SMILES C1=CC=C2C(=C1)C(=CC(=C2N)O)S(=O)(=O)O

Specifications

Melting Point ∼295°C (decomposition)
Quantity 25 g
Beilstein 2697469
Merck Index 14,454
Solubility Information Soluble in bases. Insoluble in water.
Formula Weight 239.25
Percent Purity 99%
Chemical Name or Material 4-Amino-3-hydroxy-1-naphthalenesulfonic acid
compliance-icons
Product Identifier
  • 4-Amino-3-hydroxy-1-naphthalenesulfonic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : 4-Amino-3-hydroxy-1-naphthalenesulfonicacid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.