missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-3-hydroxybenzoic acid, 98%

Product Code. p-7080613
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11499522

Brand: Thermo Scientific Alfa Aesar A10207.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2374-03-0 | C7H7NO3 | 153.137 g/mol

4-Amino-3-hydroxybenzoic acid is used in the preparation of various pharmaceutical compounds such as sphingosine kinase inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Amino-3-hydroxybenzoic acid is used in the preparation of various pharmaceutical compounds such as sphingosine kinase inhibitors.

Solubility
Soluble in chloroform, and aqeous ethanol, water (Partly miscible)

Notes
Store in cool, dry conditions in well sealed containers. keep container tightly closed. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2374-03-0
Molecular Formula C7H7NO3
Molecular Weight (g/mol) 153.137
MDL Number MFCD00017094
InChI Key NFPYJDZQOKCYIE-UHFFFAOYSA-N
Synonym 3-hydroxy-4-aminobenzoic acid, 4-amino-3-hydroxy benzoic acid, benzoic acid, 4-amino-3-hydroxy, chembl1462, 2hdr, pubchem14418, 2-amino-5-carboxyphenol, intermediates-zcf02635, acmc-209g5k, 4-carboxy-2-hydroxyaniline
PubChem CID 137566
IUPAC Name 4-amino-3-hydroxybenzoic acid
SMILES C1=CC(=C(C=C1C(=O)O)O)N

Specifications

Melting Point ∼215°C (decomposition)
Quantity 5 g
Beilstein 509859
Solubility Information Soluble in chloroform,and aqeous ethanol,water (Partly miscible)
Formula Weight 153.14
Percent Purity 98%
Chemical Name or Material 4-Amino-3-hydroxybenzoic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.