missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-3-methoxybenzoic acid, 95%

Product Code. 11347589
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11347589 5 g 5g
11337589 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11347589 Supplier Thermo Scientific Alfa Aesar Supplier No. H28098.06
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2486-69-3 | C8H9NO3 | 167.164 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2486-69-3
Molecular Formula C8H9NO3
Molecular Weight (g/mol) 167.164
MDL Number MFCD00016539
InChI Key JNFGLYJROFAOQP-UHFFFAOYSA-N
Synonym 4-amino-3-methoxy-benzoic acid, 3-methoxy-4-aminobenzoic acid, 4-amino-m-anisic acid, benzoic acid, 4-amino-3-methoxy, o-anisidine-4-carboxylic acid, 4-amino-3-methoxy benzoic acid, timtec-bb sbb008622, 4-amino-3-methoxybenzoic, 4-amino-3-methoxybenzoic acid, 4-amino-3-methoxybenzoicacid
PubChem CID 288057
IUPAC Name 4-amino-3-methoxybenzoic acid
SMILES COC1=C(C=CC(=C1)C(=O)O)N

Specifications

Melting Point 186°C to 188°C
Quantity 5 g
Formula Weight 167.16
Percent Purity 95%
Chemical Name or Material 4-Amino-3-methoxybenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.