missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-3-methylphenol, 98%

Product Code. 11416386
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11416386 25 g 25g
11426386 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11416386 Supplier Thermo Scientific Alfa Aesar Supplier No. B23152.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2835-99-6 | C7H9NO | 123.155 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2835-99-6
Molecular Formula C7H9NO
Molecular Weight (g/mol) 123.155
MDL Number MFCD00007871
InChI Key QGNGOGOOPUYKMC-UHFFFAOYSA-N
Synonym 4-amino-m-cresol, 4-hydroxy-2-methylaniline, 3-methyl-4-aminophenol, p-amino-m-cresol, phenol, 4-amino-3-methyl, p-hydroxy-o-toluidine, 2-methyl-4-hydroxyaniline, 2-amino-5-hydroxytoluene, m-cresol, 4-amino, 4-amino-3-cresol
PubChem CID 17819
ChEBI CHEBI:55546
IUPAC Name 4-amino-3-methylphenol
SMILES CC1=C(C=CC(=C1)O)N

Specifications

Melting Point 176°C to 179°C
Quantity 25 g
UN Number UN3077
Beilstein 2078803
Formula Weight 123.16
Percent Purity 98%
Chemical Name or Material 4-Amino-3-methylphenol

RUO – Research Use Only

Product Title
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.