missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-3-nitrobenzonitrile, 98%

Product Code. 11425457
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11425457 1 g 1g
11435457 5 g 5g
11445457 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11425457 Supplier Thermo Scientific Alfa Aesar Supplier No. B23463.03
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 6393-40-4 | C7H5N3O2 | 163.14 g/mol

4-Amino-3-nitrobenzonitrile is used in gamma irradiations effect on release of this compound from polyanhydride matrixes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Amino-3-nitrobenzonitrile is used in gamma irradiations effect on release of this compound from polyanhydride matrixes.

Solubility
Soluble in ethanol and benzene.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6393-40-4
Molecular Formula C7H5N3O2
Molecular Weight (g/mol) 163.14
MDL Number MFCD00013373
InChI Key JAHADAZIDZMHOP-UHFFFAOYSA-N
Synonym 3-nitro-4-aminobenzonitrile, 4-cyano-2-nitroaniline, benzonitrile, 4-amino-3-nitro, 4-amino-3-nitro-benzonitrile, 2-nitro-4-cyanoaniline, 4-amino-3-nitrobenzenecarbonitrile, zlchem 228, pubchem4669, 4-ammo-3-nitrobenzonitrile
PubChem CID 595901
IUPAC Name 4-amino-3-nitrobenzonitrile
SMILES NC1=CC=C(C=C1[N+]([O-])=O)C#N

Specifications

Melting Point 159°C to 161°C
Quantity 1 g
UN Number UN3439
Solubility Information Soluble in ethanol and benzene.
Formula Weight 163.14
Percent Purity 98%
Chemical Name or Material 4-Amino-3-nitrobenzonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.