missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-3-nitrophenol, 98%

Product Code. p-7045120
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11387446

Brand: Thermo Scientific Alfa Aesar L09487.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 610-81-1 | C6H6N2O3 | 154.125 g/mol

4-Amino-3-nitrophenol was used in the synthesis of benzimidazole based Factor Xa (trypsin-like serine protease) inhibitors and 4-(4-amino-3-nitrophenoxy)phthalonitrile. It was also employed as matrix during visible matrix-assisted laser desorption/ionization mass spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Amino-3-nitrophenol was used in the synthesis of benzimidazole based Factor Xa (trypsin-like serine protease) inhibitors and 4-(4-amino-3-nitrophenoxy)phthalonitrile. It was also employed as matrix during visible matrix-assisted laser desorption/ionization mass spectrometry.

Solubility
Soluble in water. (1.8 g/L.)

Notes
Store at room temperature. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 610-81-1
Molecular Formula C6H6N2O3
Molecular Weight (g/mol) 154.125
MDL Number MFCD00066310
InChI Key IQXUIDYRTHQTET-UHFFFAOYSA-N
Synonym 4-hydroxy-2-nitroaniline, 3-nitro-4-aminophenol, phenol, 4-amino-3-nitro, 4-amino-3-nitro-phenol, unii-r5wy41q95z, 3-nitro-4-amino phenol, 4-aminophenol-3-nitrophenol, pubchem1481, 4-amino-3-nitropbenol
PubChem CID 3758882
IUPAC Name 4-amino-3-nitrophenol
SMILES C1=CC(=C(C=C1O)[N+](=O)[O-])N

Specifications

Melting Point 148°C to 153°C
Odor Characteristic
Quantity 5 g
Beilstein 2210196
Solubility Information Soluble in water. (1.8g/L.)
Formula Weight 154.13
Percent Purity 98%
Chemical Name or Material 4-Amino-3-nitrophenol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.