missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-4'-chlorobiphenyl, 97%

Product Code. p-4380037 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11955261

Brand: Thermo Scientific Acros 444460010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 135-68-2 | C12H10ClN | 203.67 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 135-68-2
Molecular Formula C12H10ClN
Molecular Weight (g/mol) 203.67
InChI Key OREQWMWYRYXCDF-UHFFFAOYSA-N
Synonym 4-amino-4'-chlorobiphenyl, 4'-chloro-1,1'-biphenyl-4-amine, 4'-chlorobiphenyl-4-ylamine, 4'-chlorobiphenyl-4-amine, 4-amino-4'-chlorodiphenyl, 1,1'-biphenyl-4-amine,4'-chloro, unii-c8tzy038qb, 4-4-chlorophenyl aniline, 4-amino-4'-chloro-biphenyl, 4-biphenylamine, 4'-chloro
PubChem CID 8675
IUPAC Name 4-(4-chlorophenyl)aniline
SMILES C1=CC(=CC=C1C2=CC=C(C=C2)Cl)N

Specifications

Melting Point 127°C to 131°C
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Merck Index 15,43
Formula Weight 203.67
Percent Purity 97%
Chemical Name or Material 4-Amino-4'-chlorobiphenyl
compliance-icons
Product Identifier
  • 4-Amino-4'-chlorobiphenyl
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.