missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-4'-chlorodiphenyl ether, 97%

Product Code. 11474537
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
This item is not returnable. View return policy

Product Code. 11474537

Brand: Thermo Scientific Alfa Aesar B21083.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 101-79-1 | C12H10ClNO | 219.67 g/mol

It is used as an active pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an active pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 101-79-1
Molecular Formula C12H10ClNO
Molecular Weight (g/mol) 219.67
MDL Number MFCD00043925
InChI Key YTISFYMPVILQRL-UHFFFAOYSA-N
Synonym 4-4-chlorophenoxy aniline, 4-amino-4'-chlorodiphenyl ether, benzenamine, 4-4-chlorophenoxy, p-p-chlorophenoxy aniline, 4-4-chlorophenoxy benzenamine, 4-chloro-4'-aminodiphenyl ether, aniline, p-chlorophenoxy, 4-amino-4-chlorodiphenyl ether, 4'-chloro-4-aminobiphenyl ether, aniline, p-p-chlorophenoxy
PubChem CID 7578
IUPAC Name 4-(4-chlorophenoxy)aniline
SMILES NC1=CC=C(OC2=CC=C(Cl)C=C2)C=C1

Specifications

Melting Point 100°C to 103°C
Odor Odorless
Quantity 10 g
Solubility Information Insoluble in water.
Formula Weight 219.67
Percent Purity 97%
Chemical Name or Material 4-Amino-4'-chlorodiphenyl ether

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.