missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-4'-chlorodiphenyl ether, 97%

Product Code. 11484537
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11484537 50 g 50g
11474537 10 g 10g
11494537 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11484537 Supplier Thermo Scientific Alfa Aesar Supplier No. B21083.18
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 101-79-1 | C12H10ClNO | 219.67 g/mol

It is used as an active pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an active pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 101-79-1
Molecular Formula C12H10ClNO
Molecular Weight (g/mol) 219.67
MDL Number MFCD00043925
InChI Key YTISFYMPVILQRL-UHFFFAOYSA-N
Synonym 4-4-chlorophenoxy aniline, 4-amino-4'-chlorodiphenyl ether, benzenamine, 4-4-chlorophenoxy, p-p-chlorophenoxy aniline, 4-4-chlorophenoxy benzenamine, 4-chloro-4'-aminodiphenyl ether, aniline, p-chlorophenoxy, 4-amino-4-chlorodiphenyl ether, 4'-chloro-4-aminobiphenyl ether, aniline, p-p-chlorophenoxy
PubChem CID 7578
IUPAC Name 4-(4-chlorophenoxy)aniline
SMILES NC1=CC=C(OC2=CC=C(Cl)C=C2)C=C1

Specifications

Melting Point 100°C to 103°C
Odor Odorless
Quantity 50 g
Solubility Information Insoluble in water.
Formula Weight 219.67
Percent Purity 97%
Chemical Name or Material 4-Amino-4'-chlorodiphenyl ether

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.