missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-6-chloro-1,3-benzenedisulfonamide, TRC

Product Code. 17921805
Change view
Click to view available options
Quantity:
5 g
25 g
50 g
Unit Size:
25g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17921805 50 g 50g
17994105 5 g 5g
17952465 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17921805 Supplier Toronto Research Chemicals Supplier No. TRCA60337550G
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121-30-2
Molecular Formula C6 H8 Cl N3 O4 S2
Molecular Weight (g/mol) 285.73
Synonym 4-Amino-6-chlorobenzene-1,3-disulphonamide, Salamide, Hydrochlorothiazide Imp. B (EP), Hydrochlorothiazide RC A (USP), Chloraminophenamide
IUPAC Name 4-amino-6-chlorobenzene-1,3-disulfonamide
SMILES Nc1cc(Cl)c(cc1S(=O)(=O)N)S(=O)(=O)N

Specifications

Quantity 50 g
InChI Formula InChI=1S/C6H8ClN3O4S2/c7-3-1-4(8)6(16(10,13)14)2-5(3)15(9,11)12/h1-2H,8H2,(H2,9,11,12)(H2,10,13,14)
Formula Weight 284.9645
Percent Purity >95%
Chemical Name or Material 4-Amino-6-chlorobenzene-1,3-disulphonamide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.