missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-m-cresol, 99+%

Product Code. 10285780 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 g
250 g
Packaging:
Glass bottle
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10285780 50 g Glass bottle 50g
10031970 250 g Glass bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10285780 Supplier Thermo Scientific Acros Supplier No. 163200500

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2835-99-6 | C7H9NO | 123.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2835-99-6
Molecular Formula C7H9NO
Molecular Weight (g/mol) 123.15
MDL Number MFCD00007871
InChI Key QGNGOGOOPUYKMC-UHFFFAOYSA-N
Synonym 4-amino-m-cresol, 4-hydroxy-2-methylaniline, 3-methyl-4-aminophenol, p-amino-m-cresol, phenol, 4-amino-3-methyl, p-hydroxy-o-toluidine, 2-methyl-4-hydroxyaniline, 2-amino-5-hydroxytoluene, m-cresol, 4-amino, 4-amino-3-cresol
PubChem CID 17819
ChEBI CHEBI:55546
IUPAC Name 4-amino-3-methylphenol
SMILES CC1=C(C=CC(=C1)O)N

Spezifikation

CAS Min % 99.0
CAS Max % 100.0
Melting Point 176°C to 180°C
Color Brown
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula H2NC6H3(CH3)OH
Quantity 50 g
Beilstein 13, 593
Solubility Information Solubility in water: insoluble
Formula Weight 123.15
Percent Purity 99+%
Physical Form Powder
Chemical Name or Material 4-Amino-m-cresol, 99+%
Mehr anzeigen Weniger anzeigen
compliance-icons
Produktidentifikator
  • 4-Amino-m-cresol
Signalwort
  • Warning
Gefahrenkategorie
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Gefahrenhinweis
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Sicherheitshinweis
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.