missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-N,N-dimethylbenzamide, 97+%

Product Code. 11355349
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11355349 1 g 1g
11365349 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11355349 Supplier Thermo Scientific Alfa Aesar Supplier No. H50364.03
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 6331-71-1 | C9H12N2O | 164.21 g/mol

4-Amino-N,N-dimethylbenzamide is an important raw material and intermediate used in pharmaceuticals, agro based and dye stuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Amino-N,N-dimethylbenzamide is an important raw material and intermediate used in pharmaceuticals, agro based and dye stuff fields.

Solubility
Very soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6331-71-1
Molecular Formula C9H12N2O
Molecular Weight (g/mol) 164.21
MDL Number MFCD00523648
InChI Key QEPGWLBMAAEBCP-UHFFFAOYSA-N
Synonym benzamide, 4-amino-n,n-dimethyl, 4-amino-n,n-dimethyl-benzamide, n,n-dimethyl 4-aminobenzamide, 4-aminophenyl-n,n-dimethylcarboxamide, 4-dimethylcarbamyl aniline, acmc-1bgt4, 4-dimethylcarbamoyl aniline, n,n-dimethyl-p-aminobenzamide, p-amino-n,n-dimethylbenzamide, n,n-dimethyl-4-aminobenzamide
PubChem CID 235527
IUPAC Name 4-amino-N,N-dimethylbenzamide
SMILES CN(C)C(=O)C1=CC=C(N)C=C1

Specifications

Melting Point 151°C to 153°C
Quantity 1 g
Solubility Information Very soluble in water.
Formula Weight 164.21
Percent Purity ≥97%
Chemical Name or Material 4-Amino-N,N-dimethylbenzamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.