missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-N,N-dimethylbenzamide, 97+%

Product Code. p-7033280
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11355349

Brand: Thermo Scientific Alfa Aesar H50364.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6331-71-1 | C9H12N2O | 164.21 g/mol

4-Amino-N,N-dimethylbenzamide is an important raw material and intermediate used in pharmaceuticals, agro based and dye stuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Amino-N,N-dimethylbenzamide is an important raw material and intermediate used in pharmaceuticals, agro based and dye stuff fields.

Solubility
Very soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6331-71-1
Molecular Formula C9H12N2O
Molecular Weight (g/mol) 164.21
MDL Number MFCD00523648
InChI Key QEPGWLBMAAEBCP-UHFFFAOYSA-N
Synonym benzamide, 4-amino-n,n-dimethyl, 4-amino-n,n-dimethyl-benzamide, n,n-dimethyl 4-aminobenzamide, 4-aminophenyl-n,n-dimethylcarboxamide, 4-dimethylcarbamyl aniline, acmc-1bgt4, 4-dimethylcarbamoyl aniline, n,n-dimethyl-p-aminobenzamide, p-amino-n,n-dimethylbenzamide, n,n-dimethyl-4-aminobenzamide
PubChem CID 235527
IUPAC Name 4-amino-N,N-dimethylbenzamide
SMILES CN(C)C(=O)C1=CC=C(N)C=C1

Specifications

Melting Point 151°C to 153°C
Quantity 1 g
Solubility Information Very soluble in water.
Formula Weight 164.21
Percent Purity ≥97%
Chemical Name or Material 4-Amino-N,N-dimethylbenzamide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.