missing translation for 'onlineSavingsMsg'
Learn More

4-Aminobenzamide, 98+%

Product Code. 11474314
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11474314 25 g 25g
11484314 100 g 100g
11494314 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11474314 Supplier Thermo Scientific Alfa Aesar Supplier No. A12594.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 2835-68-9 | C7H8N2O | 136.154 g/mol

4-Aminobenzamide was used as a poly(ADP-ribose)polymerase (PADPRP) inhhibitor to study the death of target cells by cytotoxic effector cells using the nuclear enzyme poly(ADP-ribose)polymerase (PADPRP).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Aminobenzamide was used as a poly(ADP-ribose)polymerase (PADPRP) inhhibitor to study the death of target cells by cytotoxic effector cells using the nuclear enzyme poly(ADP-ribose)polymerase (PADPRP).

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2835-68-9
Molecular Formula C7H8N2O
Molecular Weight (g/mol) 136.154
MDL Number MFCD00007999
InChI Key QIKYZXDTTPVVAC-UHFFFAOYSA-N
Synonym p-aminobenzamide, benzamide, 4-amino, 4-amino-benzamide, p-carbamoylaniline, p-aminobenzoic acid amide, benzamide, p-amino, 4-amino benzamide, unii-77722i6pac, ccris 6792, aminobenzamide-4
PubChem CID 76079
IUPAC Name 4-aminobenzamide
SMILES C1=CC(=CC=C1C(=O)N)N

Specifications

Melting Point 182°C to 184°C
Quantity 25 g
Beilstein 2079485
Solubility Information Slightly soluble in water.
Formula Weight 136.15
Percent Purity ≥98%
Chemical Name or Material 4-Aminobenzamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.