missing translation for 'onlineSavingsMsg'
Learn More

4-Aminobenzamide, 98%

Product Code. 10601832 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10601832 25 g Glass bottle 25g
10516451 100 g Glass bottle 100g
10713095 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10601832 Supplier Thermo Scientific Acros Supplier No. 351640250

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 2835-68-9 | C7H8N2O | 136.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2835-68-9
Molecular Formula C7H8N2O
Molecular Weight (g/mol) 136.15
MDL Number MFCD00007999
InChI Key QIKYZXDTTPVVAC-UHFFFAOYSA-N
Synonym p-aminobenzamide, benzamide, 4-amino, 4-amino-benzamide, p-carbamoylaniline, p-aminobenzoic acid amide, benzamide, p-amino, 4-amino benzamide, unii-77722i6pac, ccris 6792, aminobenzamide-4
PubChem CID 76079
IUPAC Name 4-aminobenzamide
SMILES C1=CC(=CC=C1C(=O)N)N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 181.0°C to 185.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula H2NC6H4CONH2
Quantity 25 g
Solubility Information Solubility in water: soluble in water.,soluble in ethanol
Formula Weight 136.15
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 4-Aminobenzamide
Show More Show Less
compliance-icons
Product Identifier
  • 4-Aminobenzamide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.