missing translation for 'onlineSavingsMsg'
Learn More

4-Aminobenzamide, TRC

Product Code. 30136464
Change view
Click to view available options
Quantity:
5 g
10 g
50 g
Unit Size:
10g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136464 50 g 50g
30135711 10 g 10g
17973265 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136464 Supplier Toronto Research Chemicals Supplier No. TRCA58781050G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2835-68-9
Molecular Formula C7 H8 N2 O
Molecular Weight (g/mol) 136.15
Synonym Benzamide, 4-amino-, Benzamide, p-amino- (6CI,7CI,8CI), 4-Aminobenzamide, (4-(Aminocarbonyl)phenyl)amine, (4-Aminophenyl)carboxamide, 4-(Aminocarbonyl)aniline, 4-Aminobenzenecarboxamide, 4-Carbamoylaniline, 4-Carboxamidoaniline, NSC 233920, NSC 36963, SR 4329, p-Aminobenzamide, p-Aminobenzenecarboxamide, p-Aminocarbonylaniline, p-Carbamoylaniline
IUPAC Name 4-aminobenzamide
SMILES NC(=O)c1ccc(N)cc1

Specifications

Quantity 50 g
InChI Formula InChI=1S/C7H8N2O/c8-6-3-1-5(2-4-6)7(9)10/h1-4H,8H2,(H2,9,10)
Formula Weight 13.0637
Chemical Name or Material 4-Aminobenzamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.