missing translation for 'onlineSavingsMsg'
Learn More

4-Aminobenzoic Acid, 99%

Product Code. 10714861 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Glass bottle
Plastic Bottle
Plastic drum
Unit Size:
1kg
250g
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10714861 250 g Glass bottle 250g
10704861 1 kg Plastic Bottle 1kg
10013663 5 kg Plastic drum 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10714861 Supplier Thermo Scientific Acros Supplier No. 146212500
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 150-13-0 | C7H7NO2 | 137.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 150-13-0
Molecular Formula C7H7NO2
Molecular Weight (g/mol) 137.14
MDL Number MFCD00007894
InChI Key ALYNCZNDIQEVRV-UHFFFAOYSA-N
Synonym p-aminobenzoic acid, paba, para-aminobenzoic acid, vitamin bx, sunbrella, p-carboxyaniline, 4-carboxyaniline, hachemina, paraminol, benzoic acid, 4-amino
PubChem CID 978
ChEBI CHEBI:30753
IUPAC Name 4-aminobenzoic acid
SMILES NC1=CC=C(C=C1)C(O)=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 186.0°C to 189.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula H2NC6H4CO2H
Quantity 250 g
Beilstein 14, 418
Fieser 02,24
Merck Index 15, 418
Solubility Information Solubility in water: 4.7g/L (20°C). Other solubilities: soluble in glacial acetic acid,soluble in ethyl acetate,slightly soluble in benzene,121g/L etanol- 1g/50mL ether,soluble in chloroform
Formula Weight 137.14
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 4-Aminobenzoic acid
Show More Show Less
Product Identifier
  • 4-Aminobenzoic acid
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P501-Dispose of contents/container to an approved waste disposal plant.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.