missing translation for 'onlineSavingsMsg'
Learn More

4-Aminobenzoic acid, 99%

Product Code. 11450553
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11450553 250 g 250g
11359429 50 g 50g
11440553 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11450553 Supplier Thermo Scientific Alfa Aesar Supplier No. A12673.30
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 150-13-0 | C7H7NO2 | 137.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
4-Aminobenzoic acid acts as a gastric acid secretion inhibitor. It is employed as an intermediate in the synthesis of folate (Vitamin BX), azo dyes and cross linking agents. It is also useful in biomedical sector for the preparation of local anesthetic and ointments. It is widely used in industries to make pharmaceutical products, flavors and preservatives. It is actively involved as a UV-blocking ingredient in sun tan cosmetics.

Solubility
Slightly soluble in water. Soluble in methanol and diethyl ether.

Notes
Air and light sensitive. Incompatible with ferric salts and oxidizing agents. May discolor on exposure to light and air. Keep the container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 150-13-0
Molecular Formula C7H7NO2
Molecular Weight (g/mol) 137.14
MDL Number MFCD00007894
InChI Key ALYNCZNDIQEVRV-UHFFFAOYSA-N
Synonym p-aminobenzoic acid, paba, para-aminobenzoic acid, vitamin bx, sunbrella, p-carboxyaniline, 4-carboxyaniline, hachemina, paraminol, benzoic acid, 4-amino
PubChem CID 978
ChEBI CHEBI:30753
IUPAC Name 4-aminobenzoic acid
SMILES NC1=CC=C(C=C1)C(O)=O

Specifications

Melting Point ∼187°C (decomposition)
Density 1.374
Quantity 250 g
Beilstein 471605
Sensitivity Air and light sensitive
Merck Index 14,423
Solubility Information Slightly soluble in water. Soluble in methanol and diethyl ether.
Formula Weight 137.14
Percent Purity 99%
Chemical Name or Material 4-Aminobenzoic acid
Product Identifier
  • 4-Aminobenzoic acid
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: p-Aminobenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.