missing translation for 'onlineSavingsMsg'
Learn More

4-Aminobenzoic Acid, Extra Pure, SLR, Fisher Chemical

Product Code. 10010030 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10010030 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10010030 Supplier Fisher Chemical Supplier No. A/2840/48
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 150-13-0
Melting Point 186°C
Color Yellow
pH 3.5
Packaging Plastic powder jar
Molecular Formula C7H7NO2
MDL Number 7894
Quantity 100 g
Formula Weight 137.14g/mol
Physical Form Powder
Product Identifier
  • 4-Aminobenzoic acid
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P501-Dispose of contents/container to an approved waste disposal plant.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.