missing translation for 'onlineSavingsMsg'
Learn More

4-Aminobenzoic acid sodium salt, 98%

Product Code. p-7042703
Click to view available options
Quantity:
25 g
100 g
250 g
Unit Size:
100g
250g
25g
This item is not returnable. View return policy

Product Code. 15415679

Brand: Thermo Scientific Alfa Aesar J63428.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Pharmaceutical intermediate | CAS: 555-06-6 | C7H6NNaO2 | 159.12 g/mol

4-Aminobenzoic acid sodium salt is used as a Pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Aminobenzoic acid sodium salt is used as a Pharmaceutical intermediate.

Solubility
Soluble in water (50 mg/ml).

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 555-06-6
Molecular Formula C7H6NNaO2
Molecular Weight (g/mol) 159.12
MDL Number MFCD00064395
InChI Key XETSAYZRDCRPJY-UHFFFAOYSA-M
Synonym sodium 4-aminobenzoate, sodium p-aminobenzoate, 4-aminobenzoic acid sodium salt, sodium aminobenzoate, aminobenzoate sodium, antergyl, pabavit, benzoic acid, 4-amino-, monosodium salt, monosodium 4-aminobenzoate, unii-75ui7quz5j
PubChem CID 517441
IUPAC Name sodium;4-aminobenzoate
SMILES C1=CC(=CC=C1C(=O)[O-])N.[Na+]

Specifications

Melting Point >300°C
Color Yellow
Quantity 25 g
Solubility Information Soluble in water (50mg/ml).
Formula Weight 159.12
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 4-Aminobenzoic acid sodium salt
compliance-icons
Product Identifier
  • 4-Aminobenzoic acid sodium salt
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : 4-Aminobenzoic acidsodium salt

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.