missing translation for 'onlineSavingsMsg'
Learn More

4-Aminobenzophenone, 98%

Product Code. 10407670 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
10 g
25 g
Unit Size:
10g
25g
This item is not returnable. View return policy

Product Code. 10407670

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1137-41-3 | C13H11NO | 197.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1137-41-3
Molecular Formula C13H11NO
Molecular Weight (g/mol) 197.24
MDL Number MFCD00007895
InChI Key RBKHNGHPZZZJCI-UHFFFAOYSA-N
Synonym 4-aminobenzophenone, 4-aminophenyl phenyl methanone, 4-benzoylaniline, p-aminobenzophenone, methanone, 4-aminophenyl phenyl, benzophenone, 4-amino, p-benzoylaniline, usaf a-233, 4-aminophenyl-phenylmethanone, 4-aminophenyl-phenyl-methanone
PubChem CID 14346
IUPAC Name (4-aminophenyl)-phenylmethanone
SMILES NC1=CC=C(C=C1)C(=O)C1=CC=CC=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 120°C to 124°C
Color Yellow to yellow-green to beige to light grey to orange to brown
Boiling Point 246°C (17.0 hPa)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula H2NC6H4COC6H5
Quantity 25 g
Beilstein 14, 81
Solubility Information Solubility in water: hardly soluble in cold water. Other solubilities: freely soluble in alcohol and ether
Formula Weight 197.24
Percent Purity 98%
Physical Form Fine Crystalline Powder
Chemical Name or Material 4-Aminobenzophenone, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Aminobenzophenone
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.