missing translation for 'onlineSavingsMsg'
Learn More

4-Aminobenzotrifluoride, 99%

Product Code. 10256690 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 mL
50 mL
Packaging:
Glass bottle
Unit Size:
10mL
50mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10256690 10 mL Glass bottle 10mL
15710839 50 mL Glass bottle 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit 10256690 Fournisseur Thermo Scientific Acros Code fournisseur 208910100
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

CAS: 455-14-1 | C7H6F3N | 161.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 455-14-1
Molecular Formula C7H6F3N
Molecular Weight (g/mol) 161.13
MDL Number MFCD00064396
InChI Key ODGIMMLDVSWADK-UHFFFAOYSA-N
Synonym 4-trifluoromethyl aniline, 4-aminobenzotrifluoride, p-aminobenzotrifluoride, p-trifluoromethylaniline, benzenamine, 4-trifluoromethyl, 4-trifluoromethyl benzenamine, p-trifluoromethyl aniline, aniline, p-trifluoromethyl, alpha,alpha,alpha-trifluoro-p-toluidine, a,a,a-trifluoro-p-toluidine
PubChem CID 9964
ChEBI CHEBI:40750
IUPAC Name 4-(trifluoromethyl)aniline
SMILES C1=CC(=CC=C1C(F)(F)F)N

Spécification

CAS Min % 98.5
CAS Max % 100.0
Melting Point 5.0°C to 8.0°C
Color Colorless to Yellow
Density 1.2960g/mL
Boiling Point 83.0°C (12.0 mmHg)
Flash Point 86°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula H2NC6H4CF3
Refractive Index 1.4830 to 1.4850
Quantity 10 mL
Beilstein 12, III, 2151
Specific Gravity 1.296
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in ether,methanol,ethanol and,aromatics
Formula Weight 161.13
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 4-Aminobenzotrifluoride, 99%
Afficher plus Afficher moins
compliance-icons compliance-icons compliance-icons compliance-icons
Identifiant produit
  • 4-Aminobenzotrifluoride
Mention d'avertissement
  • Danger
Catégorie de danger
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 1
Mention de danger
  • H318-Causes serious eye damage.
  • H372-Causes damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H331-Toxic if inhaled.
  • H301-Toxic if swallowed.
Conseils de prudence
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.