missing translation for 'onlineSavingsMsg'
Learn More

4-Aminohippuric acid, 97%, Thermo Scientific™

Product Code. 11454854
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11454854 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11454854 Supplier Thermo Scientific Alfa Aesar Supplier No. A13786.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
4-Aminohippuric Acid is used in the measurement of renal plasma flow as a diagnostic tool which may be applied towards kidney disorders. It is used as muscle relaxant (smooth). It is also used to help differentiate and characterized organic anion transporter (OAT) isoforms.

Solubility
Soluble in alcohol, chloroform, benzene, and acetone. Insoluble in water, ether, carbon, and tetrachloride.

Notes
Store at -20°C. Light sensitive. Air sensitive. Always store away from incompatible compounds such as oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 61-78-9
Molecular Formula C9H10N2O3
Molecular Weight (g/mol) 194.19
MDL Number MFCD00007890
InChI Key HSMNQINEKMPTIC-UHFFFAOYSA-N
Synonym 4-aminohippuric acid, aminohippuric acid, p-aminohippuric acid, paha, glycine, n-4-aminobenzoyl, n-4-aminobenzoyl glycine, para-aminohippuric acid, aminohippurate, nefrotest, n-p-aminobenzoyl glycine
PubChem CID 2148
ChEBI CHEBI:104011
IUPAC Name 2-[(4-aminobenzoyl)amino]acetic acid
SMILES C1=CC(=CC=C1C(=O)NCC(=O)O)N

Specifications

Melting Point ∼200°C (decomposition)
Quantity 25 g
Beilstein 1213676
Merck Index 14,442
Formula Weight 194.19
Percent Purity 97%
Chemical Name or Material 4-Aminohippuric acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.